Orders LinkContactUsLinkComponent
VWR® Hybridization Ovens
VWR® Hybridization Ovens
Model 5420, 115 V, 60 Hz, 320 W, 33.7W x 24.8L x 24.1H cm (131/4 x 93/4 x 91/2"), 47.6W x 35.9L x 47.9H cm (183/4 x 141/8 x 187/8"), 100 °C, 5 - 60 rpm, ±0.5 °C, 24.5 kg (54 lbs.)
Catalog # 97005-252
cUL Listed
CSA Certified
Product available on GSA Advantage®
UL Listed
Supplier:   VWR International
VWR® Hybridization Ovens
Catalog # 97005-252
Supplier:  VWR International
cUL Listed
CSA Certified
Product available on GSA Advantage®
UL Listed
VWR® Hybridization Ovens
VWR® Hybridization Ovens
Model 5420, 115 V, 60 Hz, 320 W, 33.7W x 24.8L x 24.1H cm (131/4 x 93/4 x 91/2"), 47.6W x 35.9L x 47.9H cm (183/4 x 141/8 x 187/8"), 100 °C, 5 - 60 rpm, ±0.5 °C, 24.5 kg (54 lbs.)
Price
$142,841.33
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Hybridization Ovens are designed to provide rapid heat-up, excellent temperature stability, and efficient mixing for all types of hybridization and incubation applications.
  • Compact design
  • Smooth, quiet, and durable
  • VWR® Two-Year Limited Parts and Labor Warranty
Model 5430, a space-saving mini unit, holds up to four 50 mL conical tubes
Optional multi-tube carousel (47746-116) includes clips to hold twelve 1.5 mL or 2.0 mL conical tubes, six 15 mL conical tubes, or four 50 mL conical tubes. Unit heats from ambient to 70°C in 25 minutes. Models 230502V and 230502V-2 are medium-capacity units, holding up to four 35x300mm, or four 35x225mm, or eight 35x150mm, or eight 35x100mm bottles. Unit heats from ambient to 65°C in 20 minutes.
Optional multi-tube carousel (47746-114) holds forty 1.5 mL or 2.0 mL conical tubes, twelve 15 mL conical tubes, or ten 50 mL conical tubes.
Model 5420 and 5420-2 are high-capacity units, holding up to ten 35x300mm bottles or twenty 35x150mm bottles. Optional large bottle carousel (47746-110) holds four 70x220mm (750 mL) bottles or four 55x180mm (400 mL) bottles. Optional multi-tube carousel (47746-112) holds one hundred 1.5 mL or 2.0 mL conical tubes, thirty-five 15 mL conical tubes, or thirty 50 mL conical tubes. Optional microplate carousel (47746-118) holds 10 sealed standard microplates or deep well plates. Unit heats from ambient to 65°C in 30 minutes.
Note: Model 5430 oven is not recommended for use with polystyrene tubes. Plates must be sealed before use in microplate carousels.
Ordering information: Model 5420 includes two standard 35x300mm glass hybridization bottles, two spare O-rings, and a stainless steel drip pan. For Models 230502V and 5420 optional multi-tube carousels, please order clips separately. Glass bottles are supplied with blue caps and silicone O-rings for standard procedures or red caps and PTFE seals for high temperature procedures. Acrylic bottles are constructed of beta-blocking acrylic and are supplied with caps and leakproof gaskets. For 230V capability, contact your VWR sales representative.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Especificaciones
  • Max. temp.:
    100 °C
  • Description:
    Model 5420
  • Electrical:
    115 V, 60 Hz, 320 W
  • Shipping Weight:
    24.5 kg (54 lbs.)
  • Rotation Speed:
    5 - 60 rpm
  • Overall Dimensions:
    47.6W x 35.9L x 47.9H cm (183/4 x 141/8 x 187/8")
  • Chamber Dimensions:
    33.7W x 24.8L x 24.1H cm (131/4 x 93/4 x 91/2")
  • Temperature Stability:
    ±0.5 °C
  • Cat. No.:
    97005-252
  • Temperature accuracy:
    ±0.5°C
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Bottle Carousel for Model 5420
    Bottle Carousel for Model 5420
    47746-110
    Log in Para disponibilidad
    Each (1UN) - $16,048.31