Ion exchange resins, gotas, BAKER ANALYZED® IONAC® NM-60 H+/OHˉ Form, Type I 16-50 Mesh (mixed bed exchanger), J.T.Baker®
JT4631-1
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Tamaño:500 g
- Referencia:JT4631-1
- Nº de proveedor:4631-01
- Bead Size Distribution: Greater than 1.250 mm:≤5 %
- Bead Size Distribution: Less than 0.315 mm:≤2 %
- Operating capacity /mixed bed at 0.02 MOhm-cm, Eq/L:≥0.55
- Resistivity/Operating Capacity Mixed Bed test (NaCl): Resistivity after exhaustion, MOhm-cm (in minutes):≥16
- ¹ Measured to a 20,000 ohm-cm resistivity end point. The unit kgr/cf = kilograins per cubic foot.:≥16
:
:Management of Change (MOC) category = N