Glass Alcohol Burner, Grobet USA
Avoid inconvenient work stoppage for refills; the fuel level is always visible through the clear glass base
Molded facets facilitate easy handling. Wick height is adjustable. The threaded metal cap is fitted with a sealing gasket to help prevent leakage. When not in use, the snuff/storage cap reduces fuel evaporation loss. Cotton wick supplied.
: Shipped with glass reservoir, wick holder, gasket, wick, and storage cap.
Frequently Bought Together