Digital Hot Plates, Corning®
About this item
High-quality, durable hot plates have added microprocessor technology with digital displays for optimal temperature control. All units are equipped with Pyroceram® glass-ceramic tops set atop rugged housing and heating elements. The digital LED temperature display is adjustable in 5°C increments, and blinks until set temperature is reached. Microprocessor control maintains consistent and repeatable settings.
- Microprocessor maintains consistent and repeatable temperature settings from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Optional external temperature controller (89090-224) truly controls temperature inside the vessel
- Bright LED 'hot' icon lights up when top plate is over 60 °C, even when heat control is turned off
- Two piece casting design deflects spills away from electronics and includes a built in mount for an optional 457 mm support rod
- Two year manufacturer's warranty.
UL and C-UL listed. CE marked.
Corresponding magnetic hotplate stirrers (heating and stirring 89090-198 and 89090-216) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: UL and C-UL listed. CE marked.
4 Options Available Below
Product Details & Documents
High-quality, durable hot plates have added microprocessor technology with digital displays for optimal temperature control. All units are equipped with Pyroceram® glass-ceramic tops set atop rugged housing and heating elements. The digital LED temperature display is adjustable in 5°C increments, and blinks until set temperature is reached. Microprocessor control maintains consistent and repeatable settings.
- Microprocessor maintains consistent and repeatable temperature settings from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Optional external temperature controller (89090-224) truly controls temperature inside the vessel
- Bright LED 'hot' icon lights up when top plate is over 60 °C, even when heat control is turned off
- Two piece casting design deflects spills away from electronics and includes a built in mount for an optional 457 mm support rod
- Two year manufacturer's warranty.
UL and C-UL listed. CE marked.
Corresponding magnetic hotplate stirrers (heating and stirring 89090-198 and 89090-216) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: UL and C-UL listed. CE marked.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 89090-206
- 470211-560
- 470211-566
- 89090-188
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
120 V Units
Product Information
- ElectricalModelTemperature RangeTop Plate DimensionsAvailabilityPrice
- Supplier Number: 6795-400DSpecifications:cUL ListedCSA CertifiedProduct available on GSA Advantage®UL Listed
More
Specifications
Cat. No.89090-206Environmentally PreferableEnvironmental certification - Supplier Number: 6795-400DSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.470211-560Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - Supplier Number: 6795-600DSpecifications:
More
Specifications
Cat. No.470211-566Environmentally PreferableEnvironmental certification - Supplier Number: 6795-600DSpecifications:cUL ListedCSA CertifiedProduct available on GSA Advantage®UL Listed
More
Specifications
Cat. No.89090-188Environmentally PreferableEnvironmental certification