VWR® Personal-Sized Incubator
Catalog # 97025-630
Supplier: VWR International
Specifications
- Capacidad:9.2 L (0.32 cu. ft.)
- Intervalo de temperaturas:Ambient +5…70 °C
- Uniformidad de la temperatura:±0.65°C at 37°C
- Estabilidad de temperatura:±0.6°C at 37°C
- Internas, An×Pr×Al:23W x 20D x 20H cm (9 x 77/8 x 77/8")
- Peso:8.3 kg (18.4 lbs.)
- Referencia:97025-630
- Dimensiones exteriores:28.5W x 28D x 33.5H cm (111/4 x 11 x 133/16")
- Detalles eléctricos:120 V, 50/60 Hz, 0.6 A
Specifications
About this item
These personal-sized incubators are ideal for use in microbiology or hematology.
- Compact Size
- Economical
- Corrosion-Resistant Interior Chamber
- Nontoxic, Optional Thermometer
- Metal Construction
The housing and door frame are entirely metal
The top of the unit features a grommet hole that accommodates a thermometer for an accurate temperature setting. A port in the rear of the unit allows for an electrical cord to be routed through the back.
A Plexiglas window in the door provides full visibility into the corrosion-resistant interior chamber. The included shelf can be adjusted to three different levels. Heating elements are incorporated into the housing bottom and sides to ensure temperature stability and uniformity.
Accessories information: Additional shelves are available separately to increase storage capacity. The optional thermometer is non-toxic and coated in fluoropolymer resin. It offers a resolution of 1°C and is 30.5 cm (12") long with a 7.6 cm (3") immersion.
Ordering information: The incubator comes with a two-year limited warranty.