ContactUsLinkComponent
Microbiology Incubators, BF Series, BINDER
Microbiology Incubators, BF Series, BINDER
Catálogo # 80079-086
Proveedor:  BINDER INC MX
Product available on GSA Advantage®
undefined
Microbiology Incubators, BF Series, BINDER
Catálogo # 80079-086
Proveedor:  BINDER INC MX
Número de proveedor:  9010-0242
Product available on GSA Advantage®
Especificaciones
  • Rango de temperatura:
    Ambient +5…100 °C
  • Volumen:
    14 cu. ft.
  • Descripción:
    BF 400 Model
  • Referencia:
    80079-086
  • Dimensiones interiores:
    39 x 20 x 32"
  • Dimensiones exteriores:
    49 x 30 x 40"
  • Detalles eléctricos:
    115 V, 50/60 Hz, 850 W
  • Temperature Accuracy:
    ≤±0.2° at 37°C
  • Peso:
    320 lbs.
  • Nº de proveedor:
    9010-0242
Especificaciones
Acerca de este artículo
Ideal for incubation tasks with large sample quantities and short temperature recovery times in microbiological laboratories or certified laboratories.
Binder incubators feature the patented APT.Line™ preheating chamber for temperature accuracy and uniformity with adjustable fan speed 0–100%. They also have adjustable ventilation by means of a rear exhaust duct with ventilation flap and front ventilation slide. The BF incubators have a temperature range from 5º above ambient to 100ºC.
The microprocessor-controlled heating system features a LED display, integrated timer, numerous temperature and time functions, and one temperature ramp. Units also include an RS-422 interface for APT-COM™ Data Control System software.
Units come equipped with a Class 3.1 temperature safety device with visual and audible temperature alarm.
Units up to 115 liters are stackable.
Información de pedido: Supplied with two chrome-plated shelves and equipped with a Class 3.1 temperature safety device (DIN 12880).