ContactUsLinkComponent
Glas-Col® Heating Tapes
Glas-Col® Heating Tapes
Catalog # 80078-794
Supplier:  GLAS-COL
Small Business Enterprise
undefined
Glas-Col® Heating Tapes
Catalog # 80078-794
Supplier:  GLAS-COL
Supplier Number:  103AFET0.610
Small Business Enterprise
Specifications
  • Anchura:
    1.6 cm (⅝")
  • Longitud:
    1.6 cm×1.5 m (⅝"×10 ft.)
  • Referencia:
    80078-794
  • An x La:
    1.6 cm x 1.5 m (5/8" x 10')
  • Detalles eléctricos:
    230 V, 300 W
  • Nº de proveedor:
    103AFET0.610
Specifications
About this item
Heat odd lengths of glass tubes and metal pipes for both permanent and temporary applications with these heating tapes.
The tapes are ideal for use in high-moisture areas, are resistant to most laboratory environmental conditions, and will easily pass a 1000 V test for electrical insulation. Ribbon-type heating elements running lengthwise in the tape are encased in a durable glass braid and are impregnated and covered with tough silicone rubber. The glass braid and silicone rubber serve to insulate the stranded terminal leads at one end and waterproof the fittings at both ends. The attached cord has a two-blade plug and a single end termination. Temperature limit: 249 °C (480 °F).
Caution: Note: Power ratings are for tapes not covered by insulation when installed.