Orders LinkContactUsLinkComponent
Mantle/Controller Package, Series TM, Glas-Col®
Mantle/Controller Package, Series TM, Glas-Col®
Supplier:   GLAS-COL
Mantle/Controller Package, Series TM, Glas-Col®
Supplier:  GLAS-COL
Certificates
Mantle/Controller Package, Series TM, Glas-Col®
Mantle/Controller Package, Series TM, Glas-Col®
2 Options Available Below
Product Details & Documents
Glas-Col® offers TM series mantles and PowrTrol power control (see 33797-414 series) together in a special package
CSA certified.
The compact control is a solid-state, proportional voltage manual control that provides precise control of the output range, adjustable from 3–100%. Control has a lighted on/off power switch, three-wire load receptacle, and three-wire cord with molded plug. Series TM mantles feature an aluminum exterior with fabric interior, units up to 600W, 120V, 5A. Mantles withstand an internal operating temperature of 450°C (840°F).
Product Family Options
Información del producto
  • For Flask Size
    Power
    Availability
    Price
  • 250 ml
    180 W
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $22,729.94
    $22,729.94
    Each (1UN)
       $22,729.94
    Supplier Number: 100DTM102PL
  • 1000 ml
    380 W
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $25,613.38
    $25,613.38
    Each (1UN)
       $25,613.38
    Supplier Number: 100DTM108PL