VWR® Atlas Pegboards
Catalog # VWRBCP3036
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Profundidad:72"
- Anchura:6"
- Descripción:Between Column Mount Pegboard
- Referencia:VWRBCP3036
Specifications
About this item
Post-mounted epoxy pegboards are 2.5 cm (1") thick and sized to mount between standard 91.4 cm (36") and 121.9 cm (48") center-to center dimensions of Atlas uprights.
- Adjustable pegboards
- Color: Black
Pegboards are adjustable in height, allowing counters to remain clear of clutter. Pegboards feature black pegs, a stainless steel trough, and a finished back.
Ordering information: Mounting brackets VWRBCPMB are required for installation and must be purchased separately.