Orders LinkContactUsLinkComponent
Dye Pipettes, Glass, Color-Coded
Dye Pipettes, Glass, Color-Coded
Dye Pipettes, Glass, Color-Coded
Dye Pipettes, Glass, Color-Coded
Calibrated to deliver with blowout.
Made of tough borosilicate glass. Color-coded for quick, accurate identification.
Perfect for pipetting dark color dye solutions. Pipettes feature an inverse scale with volume graduations starting at zero at tip and are easier to read than conventional amber markings.
Product Family Options
  • Capacity
    Subdivisions
    Tolerance
    Color Code
  • 5 ml
    0.1 mL
    ±0.04 ml
    Blue
    Paquete de 12
    Paquete de 12 - $4,889.01
    $4,889.01
    Paquete de 12
       $4,889.01
     : PF8144K
  • 10 ml
    0.1 mL
    ±0.06 ml
    Orange
    Paquete de 12
    Paquete de 12 - $6,789.64
    $6,789.64
    Paquete de 12
       $6,789.64
     : PF8144M