Orders LinkContactUsLinkComponent
PYREX® Serological Pipettes, Wide Tip
PYREX® Serological Pipettes, Wide Tip
   Sustainable Options Available
Supplier:   Corning
PYREX® Serological Pipettes, Wide Tip
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Serological Pipettes, Wide Tip
3 Options Available Below
Product Details & Documents
Glass, reusable. These serological pipettes are made with a large tip opening to allow more rapid intake and delivery of cell suspensions and viscous materials.
  • Calibrated to deliver (TD)
  • Colored graduations are enameled on to the glass
  • Color-coded by size for easy identification and sorting
Specifications for Product Family
Specifications
  • Catalog No:
  • 89090-936
  • 53220-570
  • 53220-592
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Capacity
    Subdivisions
    Tolerance
    Availability
    Price
  • 89090-936
    PYREX® Reusable Serological Pipette with Large Tip Opening
    PYREX® Reusable Serological Pipette with Large Tip Opening
    5 ml
    0.1 ml
    ±0.1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $3,750.00
    $3,750.00
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $2,823.25
    $2,823.25
    Paquete de 6
       $3,750.00
    Case de 2 (6UN)
       $2,823.25
    Supplier Number: 7087-5
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-936
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 53220-570
    PYREX® Reusable Serological Pipette with Large Tip Opening
    PYREX® Reusable Serological Pipette with Large Tip Opening
    10 ml
    0.1 ml
    ±0.1 ml
    Log in for availability
    Log in for availability
    Case (12UN)
    Case (12UN) - $7,910.00
    $7,910.00
    Case (12UN)
       $7,910.00
    Supplier Number: 7087-10
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    53220-570
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 53220-592
    PYREX® Reusable Serological Pipette with Large Tip Opening
    PYREX® Reusable Serological Pipette with Large Tip Opening
    25 ml
    0.1 ml
    ±0.1 ml
    Log in for availability
    Log in for availability
    Case (12UN)
    Case (12UN) - $11,340.00
    $11,340.00
    Case (12UN)
       $11,340.00
    Supplier Number: 7087-25
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    53220-592
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact