PYREX® Reusable Serological Pipettes, Glass, Corning
Supplier: Corning
Certificates
About this item
Reusable serological pipettes made of PYREX® glass.
- Calibrated to deliver (TD)
- Colored graduations are enameled on to the glass
- Color-coded by size for easy identification and sorting
Tip and top are tempered to provide increased strength. Pipettes are calibrated 'to deliver' and must be blown out to obtain total rated capacity.
Certifications: ASTM E-1044
Specifications
- Catalog No:
- 89090-934
- 53204-417
- 53204-370
- 53204-359
- 53204-337
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey