Orders LinkContactUsLinkComponent
PYREX® Serological Pipettes, Corning
PYREX® Serological Pipettes, Corning
Supplier:   Corning
PYREX® Serological Pipettes, Corning
Supplier:  Corning
Certificates
PYREX® Serological Pipettes, Corning
PYREX® Serological Pipettes, Corning
2 Options Available Below
Product Details & Documents
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
  • All pipettes have negative graduations
  • Fire-polished tips for burr-free, even flow rates
Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size.
Certifications: Colors conform to I.S.O. standards.
Packaging: Non sterile pipettes available in bulk package; sterile pipettes available in individual paper-plastic wrappers or as Multi-Pack in plastic bags.
Product Family Options
Shorty, sterile, plugged, individually wrapped
Información del producto
  • Capacity
    Negative Graduations
    Subdivisions
    Packed
    Availability
    Price
  • 53284-460
    Pipet, sterile serological Pyrex®
    Pipet, sterile serological Pyrex®
    25 ml
    5.0 ml
    0.2 ml
    Individually Wrapped
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (400UN)
    Case (400UN) - $36,480.00
    $36,480.00
    Case (400UN)
       $36,480.00
    Supplier Number: 7077B-25
  • 53284-463
    Pipet, sterile serological Pyrex®
    Pipet, sterile serological Pyrex®
    50 ml
    3.0 ml
    0.5 ml
    Individually Wrapped
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (50UN)
    Case (50UN) - $8,750.00
    $8,750.00
    Case (50UN)
       $8,750.00
    Supplier Number: 7077B-50