Corning® Stripette® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
Certificates
About this item
PS, transparent, graduated, sterile.
- Calibrated to deliver (TD, Ex)
- High performance anti-drip tips, colour-coded magnifier stripe and negative graduations
- Certified RNase-/DNase-free and non pyrogenic
* Wide orifice for viscous materials.
Paper/plastic wrap allows easy opening by either pop-through or peel-apart techniques. Ideal for sterile tissue culture applications. Wrap reduces static cling.
Exclusive anti-drip tip assures accurate delivery (25, 50, and 100 ml sizes). Color-coded magnifier strips make volume reading easier. Bidirectional graduations on the pipettes provide added flexibility.
Packaging: Four packaging options:
Bulk packed in bags
Individually wrapped in plastic bags
Individually wrapped in paper-plastic bags
Individually wrapped in paper-plastic bags, triple-bagged
Bulk packed in bags
Individually wrapped in plastic bags
Individually wrapped in paper-plastic bags
Individually wrapped in paper-plastic bags, triple-bagged
Specifications
- Catalog No:
- 29443-045
- 29443-051
- 29443-049
- 29442-422
- 89089-892
- 89089-884
- 89091-046
- 29442-430
- 29442-436
- 29442-410
- 29442-418
- 29443-047
- 29442-440
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Specifications
Related Information
Certificates
Search by Lot Number Frequently Bought Together