Orders LinkContactUsLinkComponent
VWR® Pipette Pump™ Pipetting Device
VWR® Pipette Pump™ Pipetting Device
VWR® Pipette Pump™ Pipetting Device
Supplier:  VWR International
Certificates
VWR® Pipette Pump™ Pipetting Device
VWR® Pipette Pump™ Pipetting Device
4 Options Available Below
Product Details & Documents
This sturdy pipetting device accepts both disposable and standard pipettes in the flexible tapered chuck, per ASTM Standard E714-80.
  • Safe, accurate and trouble-free pipetting
  • Simple, one-handed operation
  • Color-coded for quick identification
Press plunger for rapid emptying. Resistant to acids and alkalis, they are color-coded for easy size identification. Disassemble all parts for cleaning and before autoclaving at 121 °C (250 °F) for 20 min except the chuck which should not be autoclaved.
Toothed wheel rotates forward or backward for precise filling or emptying.
Ordering information: For replacement pipettor chucks, contact your Avantor representative.
Documents
Product Family Options
Información del producto
  • Capacity
    Color
    Availability
    Price
  • 0.2 ml
    Yellow
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,641.44
    $1,641.44
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,789.17
    $1,789.17
    Each (1UN)
       $1,641.44
    Case de 12 (1UN)
       $1,789.17
  • 2 ml
    Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,116.06
    $1,116.06
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,216.51
    $1,216.51
    Each (1UN)
       $1,116.06
    Case de 12 (1UN)
       $1,216.51
  • 10 ml
    Green
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,196.01
    $1,196.01
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,303.66
    $1,303.66
    Each (1UN)
       $1,196.01
    Case de 12 (1UN)
       $1,303.66
  • 25 ml
    Red
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $827.78
    $827.78
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $902.28
    $902.28
    Each (1UN)
       $827.78
    Case de 12 (1UN)
       $902.28