Orders LinkContactUsLinkComponent
VWR® Disposable Transfer Pipettes with Reference Lines
VWR® Disposable Transfer Pipettes with Reference Lines
VWR® Disposable Transfer Pipettes with Reference Lines
VWR® Disposable Transfer Pipettes with Reference Lines
Pipettes are designed with a small tip to ensure consistent reproduction of the drop size.
  • Polyethylene construction
  • One-piece design
These one-piece polyethylene transfer pipettes feature reference lines at 1 and 2 ml.
 : Nonsterile pipettes are bulk packaged in two easy-to-use dispenser boxes of 500 each. Sterile pipettes are individually wrapped.
Product Family Options
  • Capacity
    Overall Length
    Sterility
    Graduations
    Maximum Graduation Volume
    Packed
  • 7 ml
    15.2 cm (6")
    Non sterile
    1 ml
    2 ml
    500/box, 1000/case
    Case (1,000UN)
    Case (1,000UN) - $532.07
    $532.07
    Paleta de 48 (1,000,UN)
    Paleta de 48 (1,000,UN) - $579.96
    $579.96
    Case (1,000UN)
       $532.07
    Paleta de 48 (1,000,UN)
       $579.96
  • 7 ml
    15.2 cm (6")
    Sterile
    1 ml
    2 ml
    1/bag, 500/box
    Case (500UN)
    Case (500UN) - $453.07
    $453.07
    Paleta de 48 (500UN)
    Paleta de 48 (500UN) - $493.85
    $493.85
    Case (500UN)
       $453.07
    Paleta de 48 (500UN)
       $493.85