Microcaps® Disposable Micropipettes, Drummond
Drummond microcaps are a fast, safe, clean, and accurate way to transfer liquids in micro quantities.
- Accurate (±1%)
- Dependable
- Easy to use
- Fast
They are precision-bore glass capillary tubes cut to predetermined lengths, so fill volumes are constant and precise.
Capillary action draws the fluid into the tube, and a squeeze of the bulb dispenses it. For spotting applications such as TLC, a provided spotting holder allows the liquid to be dispensed by touching the tube to the spotting surface.
Microcaps are inexpensive and disposable.
: This kit includes 100 microcaps and one bulb dispenser.
Drummond microcaps are a fast, safe, clean, and accurate way to transfer liquids in micro quantities.
- Accurate (±1%)
- Dependable
- Easy to use
- Fast
They are precision-bore glass capillary tubes cut to predetermined lengths, so fill volumes are constant and precise.
Capillary action draws the fluid into the tube, and a squeeze of the bulb dispenses it. For spotting applications such as TLC, a provided spotting holder allows the liquid to be dispensed by touching the tube to the spotting surface.
Microcaps are inexpensive and disposable.
: This kit includes 100 microcaps and one bulb dispenser.
Product Family Options
- DescriptionCapacityLengthExt.ØØ int.
- Standard Size Microcaps0.25 µl32 mm0.0210"0.0039"Paquete de 100$520.33: 1-000-00025
- Standard Size Microcaps1 µl32 mm0.0260"0.0079"-: -
- Standard Size Microcaps2 µl32 mm0.0300"0.0111"-: -
- Standard Size Microcaps3 µl32 mm0.0340"0.0136"Paquete de 100$405.01: 1-000-0030
- Standard Size Microcaps10 µl41 mm0.0315"0.0219"-: -
- Standard Size Microcaps15 µl54 mm0.0340"0.0234"Paquete de 100$405.01: 1-000-0150
- Standard Size Microcaps20 µl64 mm0.0355"0.0248"-: -
- -: -