ContactUsLinkComponent
Calibrated Micropipettes, Drummond Scientific
  53507-280
undefined
Calibrated Micropipettes, Drummond Scientific
  53507-280
 :  2-000-005
  • Capacidad:
    5 µl
  • Color:
    White
  • Descripción:
    Disposable Micropipettes
  • Referencia:
    53507-280
  • Nº de proveedor:
    2-000-005
Borosilicate glass micropipettes are ideal for a variety of liquid transfer work.
  • Color-coded micropipettes with imprinted sizes
  • Fire polished
  • Accuracy of ±0.5%
  • Anticoagulant treatment for clinical applications (special limited sizes)
The fire-polished micropipettes offer ±0.5% accuracy.
Ideal for a variety of liquid transfer work, 250 color-coded micropipettes and one 15" aspirator tube are supplied in an easy-to-store cylinder.
 : Packs include 250 micropipettes with one 38.1 cm (15") rubber aspirator tube supplied in a conveniently designed cylinder. Sizes of Precise Calibrated Micropipettes range from one to 200 µl. For blood transfer work, a heparinized 44.7 µl pipette is also available.