VWR® Disposable Micropipettes
About this item
Borosilicate glass micropipettes are made to exacting specifications and are calibrated to contain with permanent fill lines.
- Color-coded
- Borosilicate glass fabrication
- Sizes from 5 to 100 µl
- Permanent fill lines
The 44.7 µl size is treated with the anticoagulant heparin for clinical applications.
Each canister is marked with bold size designations for quick identification in the laboratory.
Ordering information: This item is packed in easy dispensing canisters of 250 micropipettes and one 38.1 cm (15") aspirator tube.
4 Options Available Below
Product Details & Documents
Borosilicate glass micropipettes are made to exacting specifications and are calibrated to contain with permanent fill lines.
- Color-coded
- Borosilicate glass fabrication
- Sizes from 5 to 100 µl
- Permanent fill lines
The 44.7 µl size is treated with the anticoagulant heparin for clinical applications.
Each canister is marked with bold size designations for quick identification in the laboratory.
Ordering information: This item is packed in easy dispensing canisters of 250 micropipettes and one 38.1 cm (15") aspirator tube.
Documents
Product Family Options
Product Information
- CapacityColorAvailabilityPrice
- 5 µlWhitePaquete de 250$1,035.02Case de 4 (250UN)$1,128.17
- 10 µlOrangePaquete de 250$1,131.62Case de 4 (250UN)$1,233.46
- 20 µlBlackPaquete de 250$1,131.62Case de 4 (250UN)$1,233.46
- 100 µlBluePaquete de 250$1,131.62Case de 4 (250UN)$1,233.46