KIMAX® Reusable Measuring (Mohr) Pipettes, Class B, Color-Coded, Kimble Chase, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Calibrated to deliver, these rugged KIMAX-51® pipettes are intended for chemical laboratory work.
- Color coding facilitates sorting and selection
- Designed from ASTM Specification E1293, Style 1, Class B requirements
Tip openings are smaller than typically desired for clinical laboratory applications. Scale is permanent brown stain fused into uniform bore tubing without etching. Pipettes are graduated to a base line which is on the straight tube above the taper.
Delivery: Each pipette is supplied with a permanently marked individual serial number and a Certificate of Graduation Accuracy.
Related Information
Frequently Bought Together