Orders LinkContactUsLinkComponent
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Pipettes are constructed of borosilicate glass and feature black enamel lettering and a graduation line.
Designed to deliver according to ASTM Specification E969, Class A requirements. Color-coded for ease in sorting and selection according to ASTM E1273. Serialized pipettes include a Certificate of Identification and Accuracy. All pipettes feature lot number and production year printed onto the pipette for QA purposes.
Product Family Options
  • Description
    Capacity
    Band Color
    Tolerance
  • Unserialized Pipets
    1 ml
    Blue
    ±0.006 ml
    Paquete de 12
    Paquete de 12 - $5,303.07
    $5,303.07
    Paquete de 12
       $5,303.07
     : 134G1
  • Unserialized Pipets
    5 ml
    White
    ±0.01 ml
    Paquete de 6
    Paquete de 6 - $2,639.87
    $2,639.87
    Paquete de 6
       $2,639.87
     : 134G5