Reusable Volumetric Pipettes, Class A, Kimble Chase
Catálogo # 75856-754
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Especificaciones
- Capacidad:10 ml
- Referencia:75856-754
- Longitud total:440 mm
- Tolerancia:±0.02 ml
- Color de la banda:Red
- Nº de proveedor:37003-10
Especificaciones
Acerca de este artículo
Calibrated to contain (bottom line) and to deliver (top line) to Class A tolerances.
The TC (to contain) feature of this pipette accommodates use with viscous liquids where the entire contents are washed out. All pipettes feature a legend of permanent brown stain and are marked with large numerals on the bulb indicating capacity. Color-coding (ASTM E1273) facilitates sorting and selection.