Orders LinkContactUsLinkComponent
Ward's® Caliche
Ward's® Caliche
Supplier:   Avantor
Ward's® Caliche
Supplier:  Avantor
Certificates
Ward's® Caliche
Ward's® Caliche
2 Options Available Below
Product Details & Documents
Hardened natural cement of calcium carbonate that binds other material such as gravel, sand, clay, and silt.
  • Collected in Arizona
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470323-126
    Caliche, bulk pack, 1 kg
    Caliche, bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $482.65
    $482.65
    Paquete de 1
       $482.65
  • 470323-116
    Caliche, student specimens
    Caliche, student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $466.90
    $466.90
    Paquete de 10
       $466.90