Orders LinkContactUsLinkComponent
MultiThief Samplers
MultiThief Samplers
Certificates
MultiThief Samplers
MultiThief Samplers
6 Options Available Below
Product Details & Documents
The MultiThief is designed for the multi-level sampling of free flowing powders and granules.
  • Variable sample volumes up to 60 ml
  • Suitable for free fl owing powders and granules
  • Can be quickly and easily changed by removing cells
Product Family Options
Información del producto
  • Material
    Length
    Ø
    Volume
    No. of Openings
    Availability
    Price
  • Sampler
    Stainless steel 316L
    1500 mm
    40 mm
    4
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $83,917.44
    $83,917.44
    Each (1UN)
       $83,917.44
    Supplier Number: 1290K-1500
  • Sampler
    Stainless steel 316
    2000 mm
    40 mm
    4
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $87,848.32
    $87,848.32
    Each (1UN)
       $87,848.32
    Supplier Number: 1290K-2000
  • Sampler
    Stainless steel 316
    1000 mm
    40 mm
    3
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $71,007.30
    $71,007.30
    Each (1UN)
       $71,007.30
    Supplier Number: 1290K-1000
Inserts
Información del producto
  • Material
    Length
    Ø
    Volume
    No. of Openings
    Availability
    Price
  • 77576-210
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    5 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18,024.01
    $18,024.01
    Each (1UN)
       $18,024.01
    Supplier Number: 1290W-05
  • 77576-214
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    25 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18,024.01
    $18,024.01
    Each (1UN)
       $18,024.01
    Supplier Number: 1290W-25
  • 77576-218
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    60 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18,024.01
    $18,024.01
    Each (1UN)
       $18,024.01
    Supplier Number: 1290W-60