Orders LinkContactUsLinkComponent
Ward's® Bacterial Form and Structure Slides
 :   Avantor
Ward's® Bacterial Form and Structure Slides
 :  Avantor
Ward's® Bacterial Form and Structure Slides
Ward's® Bacterial Form and Structure Slides
What are these things?
  • Multiple options for presentation
  • From basic forms to bacterial cell structure
  • Mixed and individual smears for the preferred presentation
The start of any microscopy lesson for bacteria is learning to identify using shape and gram stain state. These slides allow presentation in either a general presentation visualized with just a basic stain, or challenge students with a mixed sample showing both gram + and gram − organisms. Individual forms are also available with just one form per slide. Once students learn the basic bacterial cell structures, sporulation can be explored to help distinguish between different bacteria.
Product Family Options
Bacilli
  • Description
    Orientation
    Stain
  • Typical bacilli
    Smear
    Gram (+)
    Each (1UN)
    Each (1UN) - $147.78
    $147.78
    Each (1UN)
       $147.78
  • Typical bacilli -mixed
    Smear
    Gram (+/–)
    Each (1UN)
    Each (1UN) - $130.03
    $130.03
    Each (1UN)
       $130.03
  • Typical bacilli
    Smear
    Gram (–)
    Each (1UN)
    Each (1UN) - $116.28
    $116.28
    Each (1UN)
       $116.28
Mixed forms
  • Description
    Orientation
    Stain
  • Bacterial forms
    Smear
    Gram (+/–)
    Each (1UN)
    Each (1UN) - $256.03
    $256.03
    Each (1UN)
       $256.03
  • Bacterial forms
    Smear
    Fuchsin
    Each (1UN)
    Each (1UN) - $116.28
    $116.28
    Each (1UN)
       $116.28
  • Bacterial forms composite
    3 Smears
    Fuchsin
    Each (1UN)
    Each (1UN) - $134.00
    $134.00
    Each (1UN)
       $134.00
  • Bacterial forms composite
    3 Smears
    Gram (+/–)
    Each (1UN)
    Each (1UN) - $155.23
    $155.23
    Each (1UN)
       $155.23
Bacterial structure
  • Description
    Orientation
    Stain
  • Bacterial capsules
    Smear
    Congo red
    Each (1UN)
    Each (1UN) - $135.18
    $135.18
    Each (1UN)
       $135.18
Cocci
  • Description
    Orientation
    Stain
  • Typical cocci-mixed
    Smear
    Gram (+/–)
    Each (1UN)
    Each (1UN) - $136.33
    $136.33
    Each (1UN)
       $136.33
  • Typical cocci
    Smear
    Gram (–)
    Each (1UN)
    Each (1UN) - $135.18
    $135.18
    Each (1UN)
       $135.18
  • Typical cocci
    Smear
    Gram (+)
    Each (1UN)
    Each (1UN) - $181.55
    $181.55
    Each (1UN)
       $181.55
Spirillum
  • Description
    Orientation
    Stain
  • Typical spirillum
    Smear
    Gram (–)
    Each (1UN)
    Each (1UN) - $143.75
    $143.75
    Each (1UN)
       $143.75
Spores
  • Description
    Orientation
    Stain
  • Mixed spores in bacteria
    Smear
    Differential spore
    Each (1UN)
    Each (1UN) - $116.28
    $116.28
    Each (1UN)
       $116.28
  • Central spores
    Smear
    Differential spore
    Each (1UN)
    Each (1UN) - $270.90
    $270.90
    Each (1UN)
       $270.90
  • Terminal spores
    Smear
    Differential spore
    Each (1UN)
    Each (1UN) - $283.80
    $283.80
    Each (1UN)
       $283.80