Orders LinkContactUsLinkComponent
VWR® Pipettor Stand
VWR® Pipettor Stand
Transparent/blue
Catalog # 82010-756
Supplier:   VWR International
VWR® Pipettor Stand
Catalog # 82010-756
Supplier:  VWR International
VWR® Pipettor Stand
VWR® Pipettor Stand
Transparent/blue
Price
$1,478.40
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Acrylic, four-place stand provides efficient, convenient storage for most major brands of pipettors.
  • Acrylic construction
  • Holds up to four pipettors
  • Efficient, convenient storage
Upper slots are 25mm wide. Lower slots are 20mm wide.
Documents
Especificaciones
  • Color:
    Transparent/blue