ContactUsLinkComponent
VWR® Modular Heating Blocks for PCR Plates, Tubes, and Strips
VWR® Modular Heating Blocks for PCR Plates, Tubes, and Strips
Catalog # 13259-260
Supplier:  VWR International
Product available on GSA Advantage®
undefined
VWR® Modular Heating Blocks for PCR Plates, Tubes, and Strips
Catalog # 13259-260
Supplier:  VWR International
Product available on GSA Advantage®
Specifications
  • Descripción:
    Block, PCR
  • Referencia:
    13259-260
  • La x An x Al:
    15.1×9.5×5.7 cm (6×3¾×2¼")
  • Para ser utilizado con:
    96-Well Plate or 12×8 Tube Strips
  • No. of Wells:
    96
  • Well Depth:
    15.5 mm
  • Block Size:
    Double
  • Well Diameter:
    6.4 mm
Specifications
About this item
Constructed from a solid anodized aluminum block.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Tapered tube wells accept 0.2 mL tubes and are spaced for easy access and removal. Available in single and double block size.