Acerca de este artículo
Finnpipette® F1 pipettors introduce a new level of sophistication to pipetting procedures. Designed to meet the demands of modern laboratories, these lightweight devices combine standard Finnpipette® features with state-of-the-art innovations to ensure reliable and repeatable performance.
- Silent volume adjustment
- Adjustable finger rest
- Lightweight design
- Secure volume lock
- Increased air boost
- Five-year warranty
The adjustable finger rest, and lightweight design maximizes comfort without sacrificing performance. Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.