SOCOREX® ACURA® Manual 815 Single-Channel Pipettors With Tip Ejectors, Fixed Volume, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Lightweight pipettors are designed for precise, accurate pipetting of solutions, reagents, acids, serums, and other liquids, and for gel-sequencing applications.
- Dispensing is accomplished with a single plunger stroke
- Stainless steel and PVDF plunger assembly is chemically resistant
- Autoclavable to 121 °C (250 °F) without disassembly or recalibration
A differentiated overshoot function and separate push button for tip ejection reduce dead volume and prevent accidental de-tipping. The glove-friendly JustTip™ adjustable tip ejector allows the use of most pipette tips.
Ordering information: Supplied with a manufacturer’s three-year warranty, including a two-year warranty on the original calibration. The 2000 µl pipettor includes an adapter for use with a Pasteur pipet. Pipettors 83009-148, -150, -152, and -154 are supplied with polypropylene fiber filters that may be used with polypropylene tips to prevent cross-contamination.
Related Information
Frequently Bought Together