VWR® Modular Heating Sand Baths
Sand Bath for 1 Block Heater, 9.5 x 7.6 x 6.4 cm (33/4 x 3 x 21/2")
Catalog # 13259-262
Supplier: VWR International
Jump to:
Product Details & Documents
For use in heating blocks, these sand baths are ideal for irregular vessels.
- Now with Best-in-Class, Exclusive 5-Year Warranty
Stainless steel construction for superior corrosion resistance. Designed to hold sand, shot, or nonvolatile liquids. Right angle thermometers (61220-740 and 61220-742) have an accuracy rating of ± 1°C.
Documents
13259-262
Imágenes 360
VWR Laboratory Equipments
Documentación del producto
Especificaciones
- Description:Sand Bath for 1 Block Heater
- L×W×H:9.5 x 7.6 x 6.4 cm (33/4 x 3 x 21/2")
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Sand for Sand Bath, 454 g (1 lb.)