Specifications
- Esterilidad:Non sterile
- Color:Clear
- Referencia:82051-298
- Empaquetado:10/Bag
- Surface Treatment:Untreated
- Working Well Volume:10 - 130 µl
- Maximum Well Volume:138 µL
- Nº de proveedor:781101
Specifications
About this item
Microplates are manufactured from clear virgin polystyrene resin selected for high optical clarity and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
- Clear virgin polystyrene construction
- High degree of optical clarity
- Stackable
- Designed to minimize air bubble formation
- Flat bottoms and rounded tops
The MICROLON™ 600 format is available in plate 82051-264 for consistent performance in binding assays
Plate 82051-264 has a Sterility Assurance Level (SAL) of 10-3. Plates 82051-306 and -308 are supplied with lids. All plates are free of detectable RNase, DNase, human DNA, and pyrogens. They have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Wells have flat bottoms and rounded square tops (square with round corners) designed to minimize wicking and reduce air bubble formation, while increasing volume capacity and surface area. Plates are stackable.