PCR plates are molded from high performance polypropylene and feature thin walls for efficient heat transfer
Plate seals work with all Neptune™ plates for seamless integration between research and storage. PCR plate seal (89092-218) is optically clear with an aggressive seal that is perfect for reducing evaporation experienced during thermal cycling. Aluminum sealing tape (89092-220) is chemically resistant, pierceable, and ideal for PCR and sample storage. Plate sealing tape (89092-222) is non-pierceable polypropylene film. This film is not recommended for sealing raised rim plates.
Compatible with many of the latest thermocyclers on the market, these PCR plates are available in 96-well low profile, non-skirted, semi-skirted, and fully skirted versions. Refer to PCR plate compatibility chart for fit with specific thermocyclers.
Well thickness: 0.381 mm (1/64")