Orders LinkContactUsLinkComponent
Universal Fit Pipet Tips, Corning®
Universal Fit Pipet Tips, Corning®
   Opciones sostenibles disponibles
Supplier:   Corning
Universal Fit Pipet Tips, Corning®
Supplier:  Corning
   Opciones sostenibles disponibles
Certificates
Universal Fit Pipet Tips, Corning®
9 Options Available Below
Product Details & Documents
Designed to assure a reliable fit with all major brand pipettors. Beveled tip ensures full sample delivery. All tips are nonpyrogenic. All 1–200µL rack packed tips are RNase- and DNase-free.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 89089-706
  • 29442-656
  • 29442-682
  • 29442-686
  • 29442-688
  • 29442-690
  • 29442-692
  • 29442-706
  • 29442-658
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Reynosa
Especificaciones
Product Family Options
Información del producto
  • Volume
    Sterile
    Tip Color
    Packed
    Availability
    Price
  • 100 - 1000 µl
    -
    Blue
    100/hinged Rack, 10 Racks/case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $1,350.00
    $1,350.00
    Case (1,000UN)
       $1,350.00
    Supplier Number: 4713
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89089-706
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 - 200 µl
    -
    Yellow
    Bulk Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (10,000UN)
    Case (10,000UN) - $6,400.00
    $6,400.00
    Case (10,000UN)
       $6,400.00
    Supplier Number: 4845
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-656
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 - 200 µl
    -
    Natural
    Rack Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $1,265.91
    $1,265.91
    Case (960UN)
       $1,265.91
    Supplier Number: 4863
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-682
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 - 200 µl
    -
    Yellow
    Rack Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $1,310.00
    $1,310.00
    Case (960UN)
       $1,310.00
    Supplier Number: 4865
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-686
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 - 200 µl
    -
    Yellow
    Bulk Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $574.00
    $574.00
    Case (1,000UN)
       $574.00
    Supplier Number: 4866
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-688
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 - 1000 µl
    -
    Blue
    Rack Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $1,550.00
    $1,550.00
    Case (1,000UN)
       $1,550.00
    Supplier Number: 4867
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-690
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 - 1000 µl
    -
    Blue
    Bulk Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $887.00
    $887.00
    Case (1,000UN)
       $887.00
    Supplier Number: 4868
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-692
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 - 1000 µl
    +
    Blue
    Rack Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $1,820.00
    $1,820.00
    Case (1,000UN)
       $1,820.00
    Supplier Number: 9032
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-706
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 - 1000 µl
    -
    Blue
    Bulk Packed
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (10,000UN)
    Case (10,000UN) - $7,560.00
    $7,560.00
    Case (10,000UN)
       $7,560.00
    Supplier Number: 4846
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-658
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact