Orders LinkContactUsLinkComponent
Variable Transformers, STACO™
Variable transformer, 120 V, 50/60 Hz, 0 - 140 V, 10 A, 11.7×17.7×14 cm (45/8× 7×51/2 "), 4.6 kg (10.25 lbs.)
  62546-048
 :  3PN1010B
Variable Transformers, STACO™
  62546-048
Variable Transformers, STACO™
Variable Transformers, STACO™
Variable transformer, 120 V, 50/60 Hz, 0 - 140 V, 10 A, 11.7×17.7×14 cm (45/8× 7×51/2 "), 4.6 kg (10.25 lbs.)
$27,141.83
Each (1UN)
Plug-in bench-type variable transformers provide linear, efficient non-distorted control and can be used wherever variable AC voltage is required
Units feature a ventilated steel case, a 1.8m (6') three-conductor grounded line cord, fused NEMA rated receptacle, and an illuminated on/off rocker switch. Graduated dial controls output voltage from 0 to 117% of the voltage setting. Metered unit 62546-053 comes with a pivot voltmeter with ±5% full scale accuracy, conveniently located atop the enclosure for easy readout.
  • Output:
    0 - 140 V, 10 A
  • Description:
    Variable transformer
  • L×W×H:
    11.7×17.7×14 cm (45/8× 7×51/2 ")
  • Electrical:
    120 V, 50/60 Hz
  • Weight:
    4.6 kg (10.25 lbs.)