ART® 300 Non-Filter Pipette Tips, Thermo Scientific
34600-100
- Volumen:1 - 300 µl
- Longitud:5.2 cm (2")
- Referencia:34600-100
- Sterile:+
- Nº de proveedor:3771
The design of these tips ensures a reliable fit across all channels for precise tip-to-tip aspiration.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Nonsterile tips are autoclavable
Single motion loading minimizes hand fatigue. The reinforced, wide bottom tray provides extra stability and reliable tip attachment. Compatible with most single and multi-channel pipettors.
: Tips 34600-100 and -102 are packaged 96 per tray, 10 trays per pack. Tips 89413-288 and is bulk packaged.