Microtiter® 96-Well Fluorescence Microplates, Thermo Scientific
Catalog # 62403-156
Supplier: Thermo Fisher Scientific
Specifications
- Color:Black
- Descripción:Flat-Bottom
- Referencia:62403-156
- Nº de proveedor:7605
Specifications
About this item
These 96-well polystyrene microplates are designed to optimize assay performance in fluorescent applications when used with light emitting markers
Black microplates reduce background signal to improve assay sensitivity, reduce crosstalk, and avoid light piping between wells. White microplates offer higher reflectivity to enhance signal and reduce crosstalk for increased dynamic range. High-binding microplates are irradiated. Plates with U-shaped bottoms have a 0.28mL well capacity; flat-bottom wells accommodate 0.33mL.
Plates are compatible with Thermo Scientific universal microplate lids (see 62402-929 series).
Plates feature raised wells to minimize cross-contamination.