Finnpipette™ F2 Multichannel Variable Volume Pipettors, Thermo Scientific
89080-406
- Volumen:30 - 300 µl
- Descripción:12-Channel Pipettor
- Referencia:89080-406
- Incrementos:1 µl
- Exactitud:±5.0 - 1.0%
- Precisión:2.0–0.3%
- Color Code:Orange
- Nº de proveedor:4662070
Finnpipette™ F2 multichannel pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures. These pipettors combine established Finnpipette™ features with state-of-the-art innovations to create a reliable and ergonomic high-performance device.
- Fully autoclavable
- An enlarged digit display improves ease of use
- Color coding simplifies instrument selection
- Easy to service
Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision. Each lightweight device offers remarkably smooth pipetting action.
These pipettors combine established Finnpipette™ features with state-of-the-art innovations to create a reliable and ergonomic high-performance device.
: Finnpipette™ F2 devices are supplied with a five-year manufacturer's warranty.