ContactUsLinkComponent
Cliniplate™ 96-Well Microplates, Thermo Scientific
Cliniplate™ 96-Well Microplates, Thermo Scientific
Catalog # 28298-606
Product available on GSA Advantage®
undefined
Cliniplate™ 96-Well Microplates, Thermo Scientific
Catalog # 28298-606
Supplier Number:  9502867
Product available on GSA Advantage®
Specifications
  • Well Volume:
    0.45 mL
  • Color:
    Black
  • Descripción:
    Black Cliniplate™
  • Matriz:
    Polystyrene
  • Referencia:
    28298-606
  • Nº de proveedor:
    9502867
Specifications
About this item
These microplates are designed for enzyme immunoassays
Ideal for use with instrumentation adapted for 96-well plates. Cliniplates™ feature excellent optical and binding precision, uniform protein binding within and between plates, and a standard raised radius well shape to avoid cross-contamination. White and black plates are designed for assays utilizing fluorescent or luminescent markers.
Ordering information: Sterile plates are individually wrapped with lids.