ContactUsLinkComponent
Biohit Family mLINE® Multichannel Mechanical Pipettors, Variable Volume, Sartorius
Biohit Family mLINE® Multichannel Mechanical Pipettors, Variable Volume, Sartorius
Catálogo # 47745-554
undefined
Biohit Family mLINE® Multichannel Mechanical Pipettors, Variable Volume, Sartorius
Catálogo # 47745-554
Número de proveedor:  725120
Especificaciones
  • Volumen:
    1 - 10 µl
  • Referencia:
    47745-554
  • Incrementos:
    0.01 µl
  • Exactitud:
    ±4.0% at 1 µl, ±1.5% at 10 µl
  • Precisión:
    ±4.0% at 1 µL, ±1.0% at 10 µL
  • Nº de proveedor:
    725120
Especificaciones
Acerca de este artículo
Sturdy yet lightweight pipettors meet the strictest performance, ergonomic, and safety criteria. Unique loading mechanism allows tip to be loaded with constant force. This ensures the tip is correctly sealed and can be ejected with minimum force. Thermal insulation of internal components provides improved accuracy. The fast, light, stepless volume adjustment and small increments allow optimal precision.
  • Excellent ergonomics to protect from injuries with exceptionally light pipetting and tip ejection forces
  • High accuracy and precision, including in repetitive, long-lasting procedures
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Increased safety with volume lock preventing accidental volume changes while pipetting
Pipettors are easy to calibrate with the included calibration tool, which can also be used as a tube cap opener. Easy cleaning and maintenance without an opening tool. Fully autoclavable. Linear stand 80087-478 is suitable for all Biohit® mechanical and electronic pipettors.
Manufactured to ISO 9001/ISO 13485, conforms to CE/IVD standards.
Simple to clean and maintain with only three parts to disassemble. Fully autoclavable.
Unique loading mechanism allows tip to be loaded with constant force. This ensures the tip is correctly sealed and can be ejected with minimum force. Thermal insulation of internal components provides improved accuracy. The fast, light, stepless volume adjustment and small increments allow optimal precision.