Orders LinkContactUsLinkComponent
MµltiFlex™ Pipette Tips, Sorenson BioScience
MµltiFlex™ Pipette Tips, Sorenson BioScience
Supplier:   Sorenson Bioscience
MµltiFlex™ Pipette Tips, Sorenson BioScience
Certificates
MµltiFlex™ Pipette Tips, Sorenson BioScience
MµltiFlex™ Pipette Tips, Sorenson BioScience
2 Options Available Below
Product Details & Documents
Ideal for loading gels and other discrete sampling applications.
  • A 5 µl graduation provides easy volume QuikCheck
Choose rounded-end tips with a 0.57 mm Ext.Ø for SDS PAGE gels, flat-ended tips with a 0.37 mm Ext.Ø for 0.4 mm sequencing gels, or flat-ended tips with a 0.17 mm Ext.Ø for 0.2 mm sequencing gels.
The 8.3 cm (3¹/₄") tip length permits easy access to narrow tubes and sample plates.
Tips provide positive displacement accuracy using standard air displacement pipettors. They fit most popular research-grade pipettors and feature a 33 mm flexible microcapillary that allows precision pipetting in the critical 0.5 to 200 µl range.
Certifications: Certified RNase- and DNase-free.
Ordering information: All tip orders are supplied in polypropylene racks of 200 tips, with the exception of item 14229-864, which is supplied as a bulk bag of 1000 tips.
Product Family Options
Product Information
  • Tip Style
    Volume
    Availability
    Price
  • Round-ended, Ext.Ø 0.57 mm
    0.5 - 200 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 1,000
    Paquete de 1,000 - $969.56
    $969.56
    Log in for availability
    Case de 10 (1,000,UN)
    Case de 10 (1,000,UN) - $1,056.81
    $1,056.81
    Paquete de 1,000
       $969.56
    Case de 10 (1,000,UN)
       $1,056.81
    Supplier Number: 28480
  • Round-ended, Ext.Ø 0.57 mm
    0.5 - 200 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 200
    Paquete de 200 - $654.07
    $654.07
    Log in for availability
    Case de 4 (200UN)
    Case de 4 (200UN) - $712.93
    $712.93
    Paquete de 200
       $654.07
    Case de 4 (200UN)
       $712.93
    Supplier Number: 13790