Acerca de este artículo
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency. Thin wall design provides an optimal seal for single and multichannel pipettors. Tips featuring the S3 polymer eliminate sample hold up and ensure a more precise sample draw and dispense. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration.
- Trays are autoclavable
- Enabling reuse for maximum sustainability
Eco-friendly empty pipette tip trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
Thin wall design provides an optimal seal for single and multichannel pipettors. Tips featuring the S³ polymer eliminate sample hold up and ensure a more precise sample draw and dispense. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration.
Especificaciones
- Catalog No:
- 89425-660
- 89140-162
- 89140-160
- 10144-826
- 89217-470
- 89140-166
- 89140-930
- 89140-164
- 89217-468
- 89140-932
- 89140-936
- 89168-750
- 89168-754
- Environmentally Preferable:
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Sustainable Packaging, Safer Human and Environmental Health
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Certificación ambiental:
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic