Orders LinkContactUsLinkComponent
IsoTip® Pipet Tips, Corning®
   Sustainable Options Available
Supplier:   Corning
IsoTip® Pipet Tips, Corning®
Supplier:  Corning
   Sustainable Options Available
Certificates
IsoTip® Pipet Tips, Corning®
5 Options Available Below
Product Details & Documents
Designed for a variety of applications where elimination of aerosol contamination is critical, including Taq/PCR applications and radioisotope handling.
  • Exceptional durability
  • All Corning® tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
  • Sterile and certified RNase-/ DNase-free and non pyrogenic
Specifications for Product Family
Specifications
  • Catalog No:
  • 10010-364
  • 29443-272
  • 29443-276
  • 29442-616
  • 29443-278
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
  • Volume
    For Use With
    Packed
    Availability
    Price
  • 0.1 - 2 µL
    Gilson® and Other Popular Ultra Micro Pipettors
    96/Rack
    Log in for availability
    Log in for availability
    Case (960UN)
    Case (960UN) - $3,040.00
    $3,040.00
    Case (960UN)
       $3,040.00
    Supplier Number: 4801
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10010-364
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 0.1 - 10 µl
    Gilson® and Other Popular Ultra Micro Pipettors
    96/Rack
    Log in for availability
    Log in for availability
    Case (960UN)
    Case (960UN) - $3,040.00
    $3,040.00
    Case (960UN)
       $3,040.00
    Supplier Number: 4807
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29443-272
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 - 1000 µL
    Most Research-Grade Pipettors
    100/Rack
    Log in for availability
    Log in for availability
    Case (1,000UN)
    Case (1,000UN) - $3,250.00
    $3,250.00
    Case (1,000UN)
       $3,250.00
    Supplier Number: 4809
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29443-276
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 - 30 µl
    Most Research-Grade Pipettors
    96/Rack
    Log in for availability
    Log in for availability
    Case (960UN)
    Case (960UN) - $3,170.00
    $3,170.00
    Case (960UN)
       $3,170.00
    Supplier Number: 4821
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-616
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 - 200 µl
    Most Research-Grade Pipettors, Extended Length
    96/Rack
    Log in for availability
    Log in for availability
    Case (960UN)
    Case (960UN) - $3,090.00
    $3,090.00
    Case (960UN)
       $3,090.00
    Supplier Number: 4810
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    29443-278
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®