- Volumen:100 - 1000 µL
- Longitud:7.9 cm (3⅛")
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO14001 (Certificate Registration No. EMS 752603)
- Referencia:53225-753
- Estilo de la punta:Standard
- Nº de proveedor:2279
Tips ensure 100% protection against aerosol and liquid contamination.
- Pre-sterilized tips
Low retention tips reduce sample loss due to adhesion, allowing for more accurate dispensing and an increased level of reproducibility.
These tips provide a universal fit and feature the SoftFit™ design, which makes loading and ejecting easy. The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with certain Eppendorf®, Biohit®, Finnpipette®, Pipetman®, Transferpette®, and Rainin® series pipettors.