About this item
Tips ensure 100% protection against aerosol and liquid contamination.
- Pre-sterilized tips
Low retention tips reduce sample loss due to adhesion, allowing for more accurate dispensing and an increased level of reproducibility.
They feature the MicroPoint™ minimized profile design and molded reference marks at 10, 50, and 100 µl. The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor and prolongs pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with certain Eppendorf®, Oxford BenchMate®, Finnpipette®, Pipetman®, and Rainin® series pipettors.
Specifications
- Catalog No:
- 89082-066
- 89082-064
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
Specifications
Related Information
Certificates
Search by Lot NumberGuide to ART Pipet Tips
Videos
Thermo Scientific ART Universal Tips