ART® 100E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Catálogo # 72830-032
Proveedor: MOLECULAR BIO-PRODUCTS INC
Especificaciones
- Volumen:1 - 100 µL
- Longitud:5.1 cm (2")
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO14001 (Certificate Registration No. EMS 752603)
- Referencia:72830-032
- Sterile:+
- Nº de proveedor:2065E
Especificaciones
Acerca de este artículo
Tips ensure 100% protection against aerosol and liquid contamination.
- Pre-sterilized tips
- Universal design for a variety of 100 µl pipettors
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required.
Certificaciones: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Embalaje: Sterile tips are packaged 96/tray, 10 trays per pack. Non sterile tips are bulk packed.