VWR® Contour™ Apron Rails
Apron rail (adjustable; to be cut in the field), loose angle supplied, Light neutral, Steel, 49 - 60", 4"
Catalog # CFA560460-A
Supplier: VWR International
Jump to:
Product Details & Documents
These rails are only to be used between base cabinets.
- Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
- No ragged edges
- Light neutral color
The complete apron is used to create a knee space for a desk area or to create a free-standing table. Apron components are sold separately.
Documents
VWR® Steel Casework
Product Literature
Specifications
- Color:Light neutral
- Description:Apron rail (adjustable; to be cut in the field), loose angle supplied
- Height:4"
- Length:49 - 60"
- Material:Steel