Falcon® 96-Well Cell Culture Plates, Corning®
Supplier: Corning
Certificates
About this item
Polystyrene plates feature deep well design to control contamination while reducing evaporation and minimizing edge effects. Plates have serrated gripping panels for easier handling and a writing patch for plate identification. All plates except the ELISA enhanced plate include lids.
- Flat bottom, with lid
- Black wells with clear bottom
- Working volume: 25 to 340 µl
- Convenient, peel-open packaging
- Available with untreated or tissue culture-treated surface chemistries
- Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Corning® Primaria™ tissue culture treatment additionally incorporates nitrogen-containing functional groups that have been shown to improve attachment and spreading of some cell types.
Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates.
Related Information
Certificates
Search by Lot NumberCorning ELISA Workflow Guide Corning® CoolBox™ and CoolRack® Consumable Compatibility Guide Corning® CoolBox™ System for Consistent and Reproducible Cooling Corning® ELISA Microplates for Biochemical and Cell-based Applications Corning® Shopping list for returning to the bench
Product Literature
Corning Special Offer
Promotion Material
Frequently Bought Together