ContactUsLinkComponent
Spectra/Por® 2 Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Spectra/Por® 2 Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Catalog # 25218-468
Small Business Enterprise
undefined
Spectra/Por® 2 Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Catalog # 25218-468
Supplier Number:  132680
Small Business Enterprise
Specifications
  • Ø:
    28.6 mm
  • Referencia:
    25218-468
  • Flat Width:
    45 mm
  • Nº de proveedor:
    132680
Specifications
About this item
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.
This clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges, macromolecular purifications and other general applications. Possessing a broad range of chemical compatibilities, Standard RC membranes can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols, and some mild or dilute organic solvents, including DMSO.
Roll length: 15.2 m (50')