VWR® pH Electrode Arm
VWR® pH Electrode Arm
Catalog # 34105-859
Supplier: VWR International
Jump to:
Product Details & Documents
Electrode arm moves in almost any direction, holding up to three electrodes safely and securely in selected positions.
- Ideal for multiple pH readings
- Accepts all standard electrodes
- Pivots 360°, Extends 53 cm (207/8")
The arm articulates at the three points to maintain electrodes in vertical position.
Designed to accept any standard glass, reference, combination, or other electrode — even a temperature compensation probe. Weighted base and spring counter-balance permit smooth, fluid movement with superior stability.
Especificaciones
- Description:VWR® pH Electrode Arm