Specifications
- Descripción:Dry ice maker Frigimat
- Referencia:89009-208
- La x An x Al:14.4x19.4x16.5 cm (5⅝x7⅝x6½") with valve attached
- Peso al enviarse:5.2 lbs.
- Nº de proveedor:388740000
Specifications
About this item
This compact device saves the time and expense of receiving a dry ice delivery.
- Easy and safe to set up and use
- Nickel-plated "pigtail" connector
- Operates at very low pressure
- Built-in dial gauge
- Transparent polycarbonate body
Set-up and use is safe and simple
Dry ice production can be viewed through the transparent polycarbonate body that secures without clamps, screws or tools ensuring easy removal of the solid block of dry ice and disconnection from the CO2 cylinder. The lightweight body is never as cold as the dry ice, even when filled, allowing safe transport of the unit and contents when wearing appropriate gloves.
A rugged nickel-plated "pigtail" connects it to the CO2 cylinder valve which is used to regulate gas flow. Operates at very low pressure (displayed on the built-in dial gauge) and incorporates multiple pressure relief safeguards. Includes a 1.8m (6ft) nickel-plated copper connecting tube with standard CGA 320 thread coupling for easy attachment to a liquid CO2 cylinder.
Can be attached to a liquid CO2 cylinder equipped with a dip tube to produce a ready-to-use 250 to 350 gram block of dry ice in only a few minutes. Each 50 lb. cylinder makes from 10-16 blocks of dry ice.